Learn More
Heidolph™ Reax Top with Small Pick-Up Attachment
Reax Top is particularly suitable for simple mixing tasks.
Brand:  Heidolph™ 541-10000-11
Product Code. 17393010
  • 395.00 € / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 5 mm shaking orbit
  • Particularly suitable for simple mixing tasks
  • In short-term operation mode, the shaking motion is triggered by pressure on the test tube tray
  • This mode is suitable for mixing up solutions, for example
  • Persistent cell pellets can be dissolved gently but powerfully in continuous operation
  • With a speed of up to 2500 rpm and a shaking orbit of 5 mm, reliable and uniform distribution of solids, and homogenization of highly viscous media are guaranteed
Specifications
Specifications
230 V
UK Plug