Learn More
Macherey-Nagel™ Receiver Plate
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
Brand:  Macherey-Nagel™ 740687.4
 View more versions of this product
Product Code. 11902542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Receiver plate accessory
  • High-throughput applications
  • Filter frits
  • Suitable for centrifugation and vacuum
Specifications
Specifications
96-well Plate
Inserted hydrophilized filter frits of 20 μm pore size