Learn More
Macherey-Nagel™ Receiver Plate
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
Brand: Macherey-Nagel™ 740689.4
This item is not returnable.
View return policy
Description
- Receiver plate accessory
- High-throughput applications
- Filter frits
- Suitable for centrifugation and vacuum
Specifications
96-well Plate | |
Inserted hydrophilized filter frits of 50μm pore size |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur