Learn More
Heidolph™ Receiving Flask
For use with rotary evaporators
Brand:  Heidolph™ 15-300-004-11
 View more versions of this product
Product Code. 10030972
  • 1460.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With S 35/20
  • Part number 15-300-004-11: includes drain valves
    • Included as a standard for all Hei-VAP Industrial models
Specifications
Specifications
Clear
Receiving Flask
35/20
Glass
Rotary Evaporators
10 L