Learn More
Lenz Laborglasintrumente Receiving Flasks with PUR Coating
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03064073
 View more versions of this product
Product Code. 11903401
  • 191.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Especially for rotary evaporators
  • With spherical joint socket S 35
  • With highly transparent PUR coating
  • Autoclavable
  • Material: DURAN™
Specifications
Specifications
Duran Glass
2 L