Learn More
UVP Rechargeable UV Lamp
Brand:  UVP 95-0181-02
 View more versions of this product
Product Code. 12005340
  • 615.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
6 W
249 x 76 x 140 mm
249 mm
76 mm
365/254 nm
140 mm
UVSL-26P
Rechargeable UV Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products