Learn More
R&D Systems™ Recombinant Canine CCL2/MCP-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 1774-MC-025
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18118270
  • 368.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Canine CCL2/MCP-1 Protein is derived from E. coli. The Recombinant Canine CCL2/MCP-1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.8 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
403981
CCL2/JE/MCP-1
E. coli-derived canine CCL2/JE/MCP-1 protein Gln24-Pro101
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.