missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine Clusterin Protein
A cDNA sequence encoding the Clusterin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11455-FLAG-2ug
This item is not returnable.
View return policy
Specifications
442971 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS DQAVSDTELQ EMSTEGSKYI NKEIKNALKG VKQIKTLIEQ TNEERKSLLS NLEEAKKKKE DALNDTKDSE TKLKASQGVC NDTMMALWEE CKPCLKQTCM KFYARVCRSG SGLVGHQLEE FLNQSSPFYF WMNGDRIDSL LENDRQQTHA LDVMQDSFNR ASSIMDELFQ DRFFTREPQD TYHYSPFSLF QRRPFFNPKF RIARNIIPFP RFQPLNFHDM FQPFFDMIHQ AQQAMDVNLH RIPYHFPIEF PEEDNRTVCK EIRHNSTGCL KMKDQCEKCQ EILSVDCSSN NPAQVQLRQE LSNSLQIAEK FTKLYDELLQ SYQEKMFNTS SLLKQLNEQF SWVSQLANLT QSEDPFYLQV TTVGSQTSDS NVPVGFTKVV VKLFDSDPIT VMIPEAVSRN NPKFMETVAE KALQEYRQKHREE | |
Greater than 95% as determined by SDS PAGE. |
Clusterin Protein | |
Research Use Only | |
CLU | |
Canine | |
Flag Tag | |
Canine Clusterin was lyophilized from PBS, pH 7.5. |