missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Integrin alpha 4 beta 1 Protein

Brand:  R&D Systems™ 10527-A4-050

429.36 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 30000806

  • 545.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Human VCAM-1/CD106 Fc Chimera (Catalog # 862-VC ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey Integrin alpha 4 beta 1 (Catalog # 10527-A4) binds with an ED50 of 4-40 ng/mL.
Specifications

Specifications

XP_005573683.1 (Integrin alpha 4) & XP_005564991.1 (Integrin beta 1)
Bioactivity
50 μg
CD49d, IA4
Lyophilized from a 0.2 μm filtered solution in PBS.
Unconjugated
3676
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Cynomolgus Monkey
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers