missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Horse PMP2 / FABP8 Protein

A DNA sequence encoding the Equus caballus (Horse) PMP2 / FABP8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7014-200ug

 View more versions of this product

Product Code. 16077040

  • 1690.00 € / 200µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

PMP2 / FABP8 Protein
Research Use Only
PMP2
Recombinant Protein
E. coli
SNKFLGTWKL TSSENFDEYM KALGVGLGTR SLGNLAGPTV IISKSGDVIT IRTESGFKNT EISFKLGQEF EETTADNRKT KSTVTLAGGK LNQVQKWNGN ETTIKRELVD GKMVVECSMA SVVCTRIYEQ V
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Horse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers