missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Adenosine Deaminase 2/CECR1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 7518-AD-010

404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18156530

  • 529.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Adenosine Deaminase 2/CECR1 Protein is derived from CHO. The Recombinant Human Adenosine Deaminase 2/CECR1 Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in HEPES, NaCl, Glycerol.
57 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
51816
Adenosine Deaminase 2/CECR1
Chinese Hamster Ovary cell line,CHO-derived human Adenosine Deaminase 2/CECR1 protein Ile30-Lys511,with a C-terminal 6-His tag
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers