missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Adenosine Deaminase/ADA Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 7048-AD-010

404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18126620

  • 529.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Adenosine Deaminase/ADA Protein is derived from Sf 21 (baculovirus). The Recombinant Human Adenosine Deaminase/ADA Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in MOPS, NaCl and Tween™ 80.
42 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
100
Adenosine Deaminase/ADA
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Adenosine Deaminase/ADA protein Met1-Leu363,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers