missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AK2 / Adenylate kinase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) AK2 / Adenylate kinase 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5635-ec-100ug

 View more versions of this product

Product Code. 15956599

  • 505.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001186128.1
AK2 / Adenylate kinase 2 Protein
Research Use Only
AK2
Recombinant Protein
E. coli
MAPSVPAAEP EYPKGIRAVL LGPPGAGKGT QAPRLAENFC VCHLATGDML RAMVASGSEL GKKLKATMDA GKLVSDEMVV ELIEKNLETP LCKNGFLLDG FPRTVRQAEM LDDLMEKRKE KLDSVIEFSI PDSLLIRRIT GRLIHPKSGR SYHEEFNPPK EPMKDDITGE PLIRRSDDNE KALKIRLQAY HTQTTPLIEY YRKRGIHSAI DASQTPDVVF ASILAAFSKA TCKDLVMFI
Greater than 80% as determined by SDS-PAGE.
204
100 μg
Not determined.
Full Length of BC009405
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers