missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-like 3 (aa 17-220)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 4954-AN-050
254.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Angiopoietin-like 3 (aa 17-220) is derived from CHO. The Recombinant Human Angiopoietin-like 3 (aa 17-220) has been validated for the following applications: Bioactivity, Inhibition Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. | |
24.6 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
27329 | |
Angiopoietin-like Protein 3/ANGPTL3 aa 17-220 | |
Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 3/ANGPTL3 protein Ser17-Pro220,with a C-terminal 6-His tag | |
Oligomer | |
Recombinant |