Learn More
R&D Systems™ Recombinant Human Annexin A1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 3770-AN-050
Product Code. 18359287
  • 567.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Annexin A1 Protein is derived from E. coli. The Recombinant Human Annexin A1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
39.4 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
301
Annexin A1
E. coli-derived human Annexin A1 protein Met1-Asn346,with a C-terminal 6-His tag
Monomer
Recombinant
Product Suggestions