Learn More
R&D Systems™ Recombinant Human Annexin A6 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 5186-A6-050
Product Code. 18168570
  • 373.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Annexin A6 Protein is derived from E. coli. The Recombinant Human Annexin A6 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
76.5 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
309
Annexin A6
E. coli-derived human Annexin A6 protein Ala2-Asp673,with a C-terminal 6-His tag
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.