missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human beta-NGF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 256-GF-100/CF
This item is not returnable.
View return policy
Description
The Recombinant Human beta-NGF Protein is derived from NS0. The Recombinant Human beta-NGF Protein has been validated for the following applications: Bioactivity.
Specifications
Lyophilized from a 0.2μm filtered solution in Acetic Acid. | |
13.5 kDa | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
4803 | |
beta-NGF | |
Mouse myeloma cell line,NS0-derived human beta-NGF protein Ser122-Ala241 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction