missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
504.00 € - 6630.00 €
Specifications
Gene ID (Entrez) | 652 |
---|---|
Molecular Weight (g/mol) | M.W. (Observed): 19-24 kDa, reducing conditions 35-41 kDa, non-reducing conditions |
Source | Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18176280
|
R&D Systems™
314-BP-010 |
10 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
505.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18157150
|
R&D Systems™
314-BP-050 |
50 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
1265.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18196670
|
R&D Systems™
314-BP-010/CF |
10 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
504.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18340588
|
R&D Systems™
314-BP-050/CF |
50 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
1265.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18156870
|
R&D Systems™
314-BP-01M |
1 mg | Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
6630.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18793989
|
R&D Systems™
314-BP-500 |
500 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
3900.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18731388
|
R&D Systems™
314-BP-500/CF |
500 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
3900.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Measured by its ability to induce BMP responsive SEAP reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.70-7.00 ng/mL. Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 3.00-30.0 ng/mL.Specifications
652 | |
Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408 | |
Unconjugated |
M.W. (Observed): 19-24 kDa, reducing conditions 35-41 kDa, non-reducing conditions | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. |