Learn More
R&D Systems™ Recombinant Human Carbonic Anhydrase III Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 2185-CA-050
Product Code. 18330578
  • 360.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Carbonic Anhydrase III Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase III Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
31 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
761
Carbonic Anhydrase III/CA3
E. coli-derived human Carbonic Anhydrase III/CA3 protein Ala2-Lys260
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.