Learn More
R&D Systems™ Recombinant Human Caspase-10 (Mch 4) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 834-CP-010
Product Code. 18117800
  • 411.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Caspase-10 (Mch 4) Protein is derived from E. coli. The Recombinant Human Caspase-10 (Mch 4) Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in HEPES, NaCl and Sucrose.
30 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
843
Caspase-10 (Mch4)
E. coli-derived human Caspase-10 protein Val220-Ile479,with a C-terminal 6-His tag
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.