missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Caspase-8 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 705-C8-010/CF

314.46 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18116850

  • 411.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Caspase-8 Protein is derived from E. coli. The Recombinant Human Caspase-8 Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
18 kDa & 10 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
841
Caspase-8
E. coli-derived human Caspase-8 protein Ser217-Asp384 (Asp285His) (p18 subunit) & Leu385-Asp479 (p10 subunit),with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers