Learn More
R&D Systems™ Recombinant Human Cathepsin B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 953-CY-010
404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18126460
  • 529.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Cathepsin B Protein is derived from NS0. The Recombinant Human Cathepsin B Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl.
37 kDa (Pro) & 29 kDa (Mature)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
1508
Cathepsin B
Mouse myeloma cell line,NS0-derived human Cathepsin B protein Arg18-Ile339 (pro) & Phe74-Ile339 (mature),both with a C-terminal 10-His tag
Pro and Mature forms
Recombinant
Product Suggestions