Learn More
R&D Systems™ Recombinant Human CD39L1/ENTPD2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 6087-EN-010
Product Code. 18108180
  • 368.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human CD39L1/ENTPD2 Protein is derived from CHO. The Recombinant Human CD39L1/ENTPD2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl 2 and Glycerol.
48 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
954
CD39L1/ENTPD2
Chinese Hamster Ovary cell line,CHO-derived human CD39L1/ENTPD2 protein Thr29-Asp460,with a C-terminal 6-His tag
Monomer
Recombinant
Product Suggestions