missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CDO Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4384-CD-050

285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18177480

  • 362.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human CDO Protein is derived from NS0. The Recombinant Human CDO Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
100.2 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
50937
CDO
Mouse myeloma cell line,NS0-derived human CDO protein Asp26-Pro943 (Leu669Ile),with a C-terminal 6-His tag
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers