Learn More
R&D Systems™ Recombinant Human Coagulation Factor III/Tissue Factor
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 2339-PA-010
Product Code. 18118370
  • 351.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Coagulation Factor III/Tissue Factor is derived from NS0. The Recombinant Human Coagulation Factor III/Tissue Factor has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl.
26 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
2152
Coagulation Factor III/Tissue Factor
Mouse myeloma cell line,NS0-derived human Coagulation Factor III/Tissue Factor protein Gly34-Glu251,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.