Learn More
R&D Systems™ Recombinant Human Coagulation Factor Xa Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 1063-SE-010
404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18389617
  • 515.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Coagulation Factor Xa Protein is derived from Sf 21 (baculovirus). The Recombinant Human Coagulation Factor Xa Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
12 kDa & 30 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl and 5 mM CaCl 2 pH 6.0.
2159
Coagulation Factor X
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Coagulation Factor X protein Leu24-Lys488 with a C-terminal 10-His tag Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa.
Active form
Recombinant
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.