missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DBH / Dopamine beta-Hydroxylase Protein
A DNA sequence encoding the Homo sapiens (Human) DBH / Dopamine beta-Hydroxylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5912-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_000778.3 | |
DBH / Dopamine beta-Hydroxylase Protein | |
Research Use Only | |
DBH | |
Recombinant Protein | |
E. coli | |
SAPRESPLPY HIPLDPEGSL ELSWNVSYTQ EAIHFQLLVR RLKAGVLFGM SDRGELENAD LVVLWTDGDT AYFADAWSDQ KGQIHLDPQQ DYQLLQVQRT PEGLTLLFKR PFGTCDPKDY LIEDGTVHLV YGILEEPFRS LEAINGSGLQ MGLQRVQLLK PNIPEPELPS DACTMEVQAP NIQIPSQETT YWCYIKELPK GFSRHHIIKY EPIVTKGNEA LVHHMEVFQC APEMDSVPHF SGPCDSKMKP DRLNYCRHVL AAWALGAKAF YYPEEAGLAF GGPGSSRYLR LEVHYHNPLV IEGRNDSSGI RLYYTAKLRR FNAGIMELGL VYTPVMAIPP RETAFILTGY CTDKCTQLAL PPSGIHIFAS QLHTHLTGRK VVTVLVRDGR EWEIVNQDNH YSPHFQEIRM LKKVVSVHPG DVLITSCTYN TEDRELATVG GFGILEEMCV NYVHYYPQTQ LELCKSAVDA GFLQKYFHLI NRFNNEDVCT CPQASVSQQF TSVPWNSFNR DVLKALYSFA PISMHCNKSS AVRFQGEWNL QPLPKVISTL EEPTPQCPTS QGRSPAGPTV VSIGGGKG | |
Greater than 90% as determined by SDS-PAGE. |
1621 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction