Learn More
R&D Systems™ Recombinant Human Dopa Decarboxylase/DDC Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 3564-DC-010
Product Code. 18364997
  • 347.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Dopa Decarboxylase/DDC Protein is derived from Sf 21 (baculovirus). The Recombinant Human Dopa Decarboxylase/DDC Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in MES, NaCl and DTT.
55 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
1644
Dopa Decarboxylase/DDC
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Dopa Decarboxylase/DDC protein Met1-Glu480,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.