Learn More
R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (157 aa) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 234-FSE-025
Product Code. 18344767
  • 399.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Human FGF basic/FGF2 (157 aa) Protein is derived from E. coli. The Recombinant Human FGF basic/FGF2 (157 aa) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
17.4 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Recombinant
2247
FGF basic 157 aa
E. coli-derived human FGF basic/FGF2/bFGF protein Gly132-Ser288
<0.1 EU per 1μg of the protein by the LAL method.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.