missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Fibronectin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 4305-FNB-200
This item is not returnable.
View return policy
Description
The Recombinant Human Fibronectin Protein is derived from NS0. The Recombinant Human Fibronectin Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
CAE45932 | |
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and Tween™ | |
207 kDa | |
Mouse myeloma cell line,NS0-derived human Fibronectin protein Gln32-Pro1908,with a C-terminal 6-His tag | |
RUO | |
0.5 to 50μg/mL | |
Unconjugated | |
Recombinant |
Bioactivity | |
2335 | |
200 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. | |
<0.10 EU per 1μg of the protein by the LAL method | |
Recombinant protein | |
Human | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction