missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Fc (HEK293) Protein

331.00 € - 5860.00 €

Specifications

Accession Number AAA17999.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) M.W. Observed: 57-64 kDa, under reducing conditions., M.W. Theoretical: 45 kDa
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18767086
View Documents
R&D Systems™
11309-FKH-010
10 μg
331.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
18738206
View Documents
R&D Systems™
11309-FKH-01M
1 mg
5860.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
18736656
View Documents
R&D Systems™
11309-FKH-050
50 μg
740.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
18795576
View Documents
R&D Systems™
11309-FKH-100
100 μg
1080.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
18709176
View Documents
R&D Systems™
11309-FKH-250
250 μg
2065.00 €
250µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.300-4.80 ng/mL.
Specifications

Specifications

AAA17999.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W. Observed: 57-64 kDa, under reducing conditions., M.W. Theoretical: 45 kDa
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Flt-3 Ligand/FLT3L
Bioactivity
2323
Human embryonic kidney cell, HEK293-derived human Flt-3 Ligand/FLT3L protein Human Flt-3 Ligand/FLT3L (Thr27-Pro185) (N-terminus) Accession # AAA17999.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
<0.10 EU per 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Videos
SDS
Documents

Documents

Product Certifications