Learn More
R&D Systems™ Recombinant Human FSH alpha/beta Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 5925-FS-010/CF
 View more versions of this product
Product Code. 18328377
  • 524.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human FSH alpha/beta Protein is derived from CHO. The Recombinant Human FSH alpha/beta Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE with silver staining.
2488
FSH alpha/beta
Chinese Hamster Ovary cell line,CHO-derived human FSH alpha/beta protein, (N-terminus) Human FSH alpha Ala25-Ser116 Accession &Num; P01215 Human FSH beta Cys21-Glu129 Accession &Num; P01225 (C-terminus)
Noncovalently-linked heterodimer
Recombinant
Product Suggestions