Learn More
R&D Systems™ Recombinant Human HAI-1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Brand:  R&D Systems™ 1048-PI-010
404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18328747
  • 529.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human HAI-1 Protein is derived from NS0. The Recombinant Human HAI-1 Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
Inhibition Assay
6692
HAI-1
Mouse myeloma cell line,NS0-derived human HAI-1 protein Pro37-Glu449,with a C-terminal 10-His tag
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
47 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions