missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human HAI-2A Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Brand:  R&D Systems™ 1106-PI-010

345.56 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18187550

  • 445.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human HAI-2A Protein is derived from NS0. The Recombinant Human HAI-2A Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
21 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
10653
HAI-2A
Mouse myeloma cell line,NS0-derived human HAI-2A protein Met1-Lys197,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers