missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HERPUD1 His Protein
Highly purified. Generating reliable and reproducible results.
394.00 € - 1070.00 €
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffer saline (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 9709 |
Molecular Weight (g/mol) | TMW: 31.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18214290
|
Novus Biologicals™
NBP2-51862-0.05MG |
0.05 mg |
394.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18243630
|
Novus Biologicals™
NBP2-51862-0.25MG |
0.25 mg |
1070.00 €
0.25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-263 of Human HERPUD1 The Recombinant Human HERPUD1 Protein is derived from E. coli. The Recombinant Human HERPUD1 Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate buffer saline (pH 7.4) containing 10% glycerol | |
TMW: 31.6kDa | |
HERPMIF1, homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-likedomain member 1 protein, homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 1, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 1 protein, KIAA0025MMS-inducible, Methyl methanesulfonate (MMF)-inducible fragment protein 1, Mif1, SUP | |
Unconjugated | |
>85%, by SDS-PAGE |
SDS-PAGE | |
9709 | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
HERPUD1 | |
Recombinant |
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human HERPUD1 His Protein