missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGF (NS0-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
970.00 € - 1645.00 €
Specifications
Accession Number | P14210.2 |
---|---|
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
Gene ID (Entrez) | 3082 |
Molecular Weight (g/mol) | alpha chain: 53.7 kDa beta chain: 26 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757736
|
R&D Systems™
294-HGN-100 |
100 μg |
970.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18178230
|
R&D Systems™
294-HGN-250 |
250 μg |
1645.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human HGF (NS0-expressed) Protein is derived from NS0. The Recombinant Human HGF (NS0-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
P14210.2 | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. | |
alpha chain: 53.7 kDa beta chain: 26 kDa | |
RUO | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Unconjugated | |
Human | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
3082 | |
Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
HGF | |
HGF | |
Reconstitute at 50 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |