Learn More
R&D Systems™ Recombinant Human His6-alpha-Synuclein Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ SP-480-500
360.25 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18694749
  • 471.00 € / 500µg
Explore more special offers
Description
Description
The Recombinant Human His6-Synuclein-alpha Protein is derived from E. coli. The Recombinant Human His6-Synuclein-alpha Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
P37840.1
6622
His6 Synuclein-alpha
E. coli-derived human alpha-Synuclein protein Met1 - Ala140 with a C-terminal 6-His tag
Unconjugated
Supplied as a 0.2 μm filtered solution in HEPES and NaCl.
15 kDa
500μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Recombinant
Product Suggestions