missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human His6-USP1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ E-564-050
This item is not returnable.
View return policy
Description
Recombinant Human USP1 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP1 concentration of 0.01-0.5 μM.
Specifications
O94782 | |
X mg/mL (X μM) in 50 mM HEPES, pH 7.5, 100 mM NaCl, 10% glycerol, 1 mM TCEP | |
M.W. (theoretical): 89 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
Unconjugated | |
>80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
Bioactivity | |
7398 | |
His6 USP1 | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP1 protein Contains substitutions at G670A & G671A and a C-terminal 6-His tag | |
USP1 | |
Recombinant | |
USP1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction