Learn More
R&D Systems™ Recombinant Human Histone Deacetylase 8 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 4359-DA-050
Product Code. 18187040
  • 362.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Histone Deacetylase 8 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Histone Deacetylase 8 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol.
43 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
55869
Histone Deacetylase 8/HDAC8
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Histone Deacetylase 8/HDAC8 protein Met1-Val377,with an N-terminal Met and 7-His tag
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.