missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IGFALS/ALS Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 9917-IA-050

365.43 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18128170

  • 478.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Derived from CHO
  • Validated for the following applications: Bioactivity.
Specifications

Specifications

P35858-1
3483
50 μg
12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution.
ACLSD, ALS, Igfals
Human
Lyophilized from a 0.2 μm filtered solution in PBS
Bioactivity
76 to 88 kDa, reducing conditions
Chinese Hamster Ovary cell line, CHO-derived human IGFALS/ALS protein Ala28-Cys605, with a C-terminal 6-His tag
< 0.10 EU per 1 μg of the protein by the LAL method
Unconjugated
Reconstitute at 500 μg/mL in PBS
> 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers