missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-18/IL-1F4 Biotinylated Protein

Biotinylated

Brand:  R&D Systems™ BT9124-050

437.14 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18726286

  • 572.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Human IL-18 BPa His-tag Protein (Catalog # 11236-BP ) is immobilized at 0.25 μg/mL (100 μL/well), the concentration of Biotinylated Recombinant Human IL-18/IL-1F4 (Catalog # BT9124) that produces 50% of the binding response is 1.50-12.0 ng/mL.
Specifications

Specifications

Q14116
Lyophilized from a 0.2 μm filtered solution in PBS and DTT with Trehalose.
M.W. Observed: 15-21 kDa, under reducing conditions., M.W. Theoretical: 18 kDa
E. coli-derived human IL-18/IL-1F4 protein Tyr37-Asp193
<0.10 EU per 1 μg of the protein by the LAL method.
Biotin
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Bioactivity
3606
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Reconstitute at 250 μg/mL in PBS.
IL-18/IL-1F4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers