missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-22 (HEK293-expressed) Protein

Brand:  R&D Systems™ 11311-IL-010

308.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18708996

  • 403.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to induce IL-10 secretion in COLO 205 human colorectal adenocarcinoma cells. Nagalakshmi, M.L. et al. (2004) International Immunopharmacology 4:679. The ED50 for this effect is 0.160-1.60 ng/mL.
Specifications

Specifications

Q9GZX6.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa
Human embryonic kidney cell, HEK293-derived human IL-22 protein Ala34-Ile179
<0.10 EU per 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Bioactivity
50616
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
IL-22
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers