missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-22 (HEK293-expressed) Protein

403.00 € - 9830.00 €

Specifications

Accession Number Q9GZX6.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 50616
Molecular Weight (g/mol) M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18708996
View Documents
R&D Systems™
11311-IL-010
10 μg
403.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
18778806
View Documents
R&D Systems™
11311-IL-250
250 μg
3390.00 €
250µg
Please to purchase this item. Need a web account? Register with us today!
18759706
View Documents
R&D Systems™
11311-IL-01M
1 mg
9830.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
18755766
View Documents
R&D Systems™
11311-IL-050
50 μg
1110.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Measured by its ability to induce IL-10 secretion in COLO 205 human colorectal adenocarcinoma cells. Nagalakshmi, M.L. et al. (2004) International Immunopharmacology 4:679. The ED50 for this effect is 0.160-1.60 ng/mL.
Specifications

Specifications

Q9GZX6.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Bioactivity
50616
Human embryonic kidney cell, HEK293-derived human IL-22 protein Ala34-Ile179
Unconjugated
IL-22
Videos
SDS
Documents

Documents

Product Certifications