Learn More
R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 6995-IL-010
 View more versions of this product
Product Code. 18156950
  • 478.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein.
17 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 100μg/mL in Water.
27179
IL-36 alpha/IL-1F6 (aa 6-158)
E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158
Monomer
Recombinant
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.