missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Insulin Protein
A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10732-100mg
This item is not returnable.
View return policy
Specifications
3630 | |
100 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
27.5 units/mg. Insulin has been evaluated in cell culture (Human Foreskin Fibroblasts). The effective concentration range for use in defined media is 1-5ug/ml. However, the optimum concentration range for a particular application must be determined by the investigator. | |
Human | |
Untagged | |
The protein was lyophilized from 50mM Sodium Chloride solution. |
Insulin Protein | |
Research Use Only | |
INS | |
Recombinant Protein | |
E. coli | |
MALWMRLLPL LALLALWGPD PAAAFVNQHL CGSHLVEALY LVCGERGFFY TPKTRREAED LQVGQVELGG GPGAGSLQPL ALEGSLQKRG IVEQCCTSIC SLYQLENYCN | |
Greater than 95.0% as determined by SDS-PAGE. |