Learn More
R&D Systems™ Recombinant Human Integrin alpha 11 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 6357-AB-050
457.01 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18158210
  • 598.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Integrin alpha 11 beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha 11 beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
Integrin alpha 11 beta 1
Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 11 beta 1 protein, (N-terminus) Human Integrin alpha 11 (Phe23-Pro1141) (Leu524Arg) Accession &Num; Q9UKX5 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus)
Noncovalently-linked heterodimer
Recombinant
134.6 kDa ( alpha11) & 88.4 (beta1)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.