missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha 4 beta 7 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 5397-A3-050

421.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18196390

  • 559.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Integrin alpha 4 beta 7 Protein is derived from CHO. The Recombinant Human Integrin alpha 4 beta 7 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and MgCl 2 .
M.W. Observed: 130-155 kDa and 100-130 kDa, under reducing conditions.
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
3676
Integrin alpha 4 beta 7/LPAM-1
Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 4 beta 7/LPAM-1 protein, (N-terminus) Human Integrin alpha 4 (Tyr34-Gln970 (R591L & R878Q)) Accession &Num; P13612 Acidic tail HHHHHH Human Integrin beta 7 (Glu20-His723) Accession &Num; P26010 Basic tail (C-terminus)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers