missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha 6 (X1) beta 4 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 5497-A6-050

420.72 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18118170

  • 534.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Mouse Laminin I (Catalog # 3400-010-02 ) is coated at 10 μg/mL, Recombinant Human Integrin alpha 6 beta 4 binds with an apparent KD <5 nM.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
104 kDa ( alpha6) & 84.8 kDa (beta4)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
3655
Integrin alpha 6 beta 4 X1 isoform
Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 6 beta 4 protein, (N-terminus) Human Integrin alpha 6 (Phe24-Lys878) Accession &Num; NP_000201 Acidic Tail HHHHHH Human Integrin beta 4 (Asn28-Ser710) Accession &Num; P16144 Basic Tail (C-terminus)
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers