missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha L beta 2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 3868-AV-050

464.78 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18107080

  • 607.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Integrin alpha L beta 2 Protein is derived from CHO. The Recombinant Human Integrin alpha L beta 2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
121.7 kDa (Integrin alphaL), 79 kDa (Integrin beta2)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
3683
Integrin alpha L beta 2
Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha L beta 2 protein, (N-terminus) Human Integrin alpha L (Tyr26-Met1089) Accession &Num; P20701 acidic tail Human Integrin beta 2 (Gln23-Asn700) Accession &Num; P05107 basic tail (C-terminus)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers