missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ISG15 AMC Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ UL-553-050

347.29 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18750599

  • 454.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This fluorogenic substrate for ISG15 hydrolases is based on the carboxy-terminus derivatization of ISG15 with 7-amido-4-methylcoumarin (AMC). Release of AMC fluorescence can be monitored with an excitation wavelength of 380 nm and an emission wavelength of 460 nm. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human ISG15 AMC concentration of 0.1-1 µM.
Specifications

Specifications

P05161.5
9636
ISG15/UCRP AMC
E. coli-derived human ISG15/UCRP protein Met1 - Gly156 (C78S, G157C) with a C-terminal 6-His tag; C-terminal AMC derivatized protein
7-Amino-4-Methyl Coumarin
Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and DTT.
M.W. Theroretical: 18 kDa
50μg
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers